Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HEPES Sodium Salt, ≥98% (dry basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
75277-39-3
C8H17N2NaO4S
260.284
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
hepes sodium salt,hepes hemisodium salt,sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate,sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate,unii-z9fto91o8a,1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt,z9fto91o8a,sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate,4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt,4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
2724248
CHEBI:46758
sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
C1CN(CCN1CCO)CCS(=O)(=O)[O-].[Na+]
75277-39-3
C8H17N2NaO4S
260.284
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
hepes sodium salt,hepes hemisodium salt,sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate,sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate,unii-z9fto91o8a,1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt,z9fto91o8a,sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate,4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt,4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
2724248
CHEBI:46758
sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
C1CN(CCN1CCO)CCS(=O)(=O)[O-].[Na+]
Linear Formula HO(CH2)2(C4H8N2)(CH2)2SO3Na
Formula Weight 260.29
Physical Form Crystalline Powder
Percent Purity ≥98%
Packaging Plastic bottle
Quantity 100g
Chemical Name or Material HEPES Sodium Salt
Assay Dry basis