Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES Sodium Salt, CellPURE™, Fisher BioReagents

Click to view available options
Quantity:
100g
Packaging:
Poly Bottle
75277-39-3
C8H17N2NaO4S
260.28
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
2724248
CHEBI:46758
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
75277-39-3
C8H17N2NaO4S
260.28
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
2724248
CHEBI:46758
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Purity 99.5% min.
Absorbance 0.05 AU max. at 260nm, 0.05 AU max. at 280nm, 0.05 AU max. at 290nm
Cell Culture Test Pass Test
Assay Percent Range ≥99.5 %
Endotoxin 0.1 EU/mg max.
Heavy Metals (as Pb) 5ppm max.
Formula Weight 260.29
Physical Form Powder/Solid
Grade Cell Culture
Infrared Spectrum Conforms to reference
Iron (Fe) 5ppm max.
Packaging Poly Bottle
Standard Plate Count 100 CFU/g max.
Water 3% Max.
RNase, DNase, Nickase, and Protease Not Detected Detected/Not Detect.
Color White
Quantity 100g
Chemical Name or Material HEPES Sodium Salt, CellPURE™, Cell culture tested
Show More Show Less

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1