Learn More
Novus Biologicals™ HEPN1 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP255007PEP
Catalog No. NBP55007PEP
Add to cart
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human HEPN1. The HEPN1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Antibody Competition
641654
>80% by SDS-PAGE and Coomassie blue staining
E. coli
RUO
HEPN1
PBS and 1M Urea, pH 7.4.
27 kDa
100 μl
Store at -20°C. Avoid freeze-thaw cycles.
HEPN1
Human