Learn More
HepPar-1 Monoclonal Antibody (OCH1E5), Zeta
Mouse Monoclonal Antibody Cocktail
$200.94
Specifications
Antigen | HepPar-1 |
---|---|
Clone | OCH1E5 |
Concentration | 200 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5875
![]() |
ZETA CORPORATION
Z2087MS |
500 μL |
Each of 1 for $200.94
|
|
|||||
Description
A recommended positive control tissue for this product is HCC, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
HepPar-1 Monoclonal antibody specifically detects HepPar-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HepPar-1 | |
200 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
tris with BSA, NP-40 and <0.1% sodium azide | |
Formalin fixed failed human allograft liver that was mechanically disrupted actual price paid for the product | |
Primary | |
2-8°C |
OCH1E5 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
HepPar-1 | |
IgG1 κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.