Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ HEPPSO, 0.2M buffer soln., pH 8.0

Click to view available options
Quantity:
100mL
250mL
68399-78-0
C9H20N2O5S
268.328
MFCD00038351
GIZQLVPDAOBAFN-UHFFFAOYSA-N
100205
CHEBI:32951
2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C1CN(CCN1CCO)CC(CS(=O)(=O)O)O
68399-78-0
C9H20N2O5S
268.328
MFCD00038351
GIZQLVPDAOBAFN-UHFFFAOYSA-N
100205
CHEBI:32951
2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C1CN(CCN1CCO)CC(CS(=O)(=O)O)O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 8.0
Quantity 250mL
Chemical Name or Material HEPPSO buffer soln.