Alfa Aesar™ Heptadecafluorodecahydro(trifluoromethyl)naphthalene

Manufacturer: Alfa Aesar™ 1615114

Quantity 25g

 View more versions of this product

Catalog No. AA1615114

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 51294-16-7
    Formula Weight 512.11
    MDL Number MFCD00014326
    Synonym Trifluoromethyl(perfluorodecahydronaphthalene); Perfluoro(methyldecalin)
    Chemical Name or Material Heptadecafluorodecahydro(trifluoromethyl)naphthalene

    Specifications

    Formula Weight 512.11
    MDL Number MFCD00014326
    CAS 51294-16-7
    Physical Form Liquid
    Refractive Index 1.3184
    Synonym Trifluoromethyl(perfluorodecahydronaphthalene); Perfluoro(methyldecalin)
    Density 1.908g/mL
    Boiling Point 140° to 142°C
    Quantity 25g
    Chemical Name or Material Heptadecafluorodecahydro(trifluoromethyl)naphthalene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit