Alfa Aesar™ Heptafluorobutyric acid, 99%

Manufacturer: Alfa Aesar™ A1490736

Quantity 500g

 View more versions of this product

Catalog No. AAA1490736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 375-22-4
    Molecular Formula C4HF7O2
    Linear Formula CF3CF2CF2CO2H
    Formula Weight 214.04
    MDL Number MFCD00004171
    UN Number UN3265
    Beilstein 1426882
    Synonym Perfluorobutyric acid
    Chemical Name or Material Heptafluorobutyric acid

    Specifications

    Formula Weight 214.04
    UN Number UN3265
    Odor Unpleasant
    Melting Point −18°C
    TSCA TSCA
    Molecular Formula C4HF7O2
    Linear Formula CF3CF2CF2CO2H
    MDL Number MFCD00004171
    CAS 375-22-4
    Beilstein 1426882
    Synonym Perfluorobutyric acid
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number ET4025000
    Boiling Point 120° to 121°C
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Heptafluorobutyric acid
    Assay 99%

    Description

    Applications
    Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

    Solubility
    Miscible with water, toluene, ether, ethanol and benzene.       

    Notes
    Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents..   

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit