Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyric acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
375-22-4
C4HF7O2
214.04
MFCD00004171
YPJUNDFVDDCYIH-UHFFFAOYSA-N
heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech
9777
CHEBI:39426
2,2,3,3,4,4,4-heptafluorobutanoic acid
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
375-22-4
C4HF7O2
214.04
MFCD00004171
YPJUNDFVDDCYIH-UHFFFAOYSA-N
heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech
9777
CHEBI:39426
2,2,3,3,4,4,4-heptafluorobutanoic acid
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
Infrared Spectrum Authentic
Melting Point -17.5°C
Refractive Index 1.3
Quantity 25g
Color Colorless to Yellow
Boiling Point 120.0°C (755.0 mmHg)
Density 1.6400g/mL
Specific Gravity 1.64
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula CF3CF2CF2CO2H
Solubility Information Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
Formula Weight 214.04
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Heptafluorobutyric acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 206-786-3