Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyric Acid 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
375-22-4
C4HF7O2
214.039
MFCD00004171
YPJUNDFVDDCYIH-UHFFFAOYSA-N
heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech
9777
CHEBI:39426
2,2,3,3,4,4,4-heptafluorobutanoic acid
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
375-22-4
C4HF7O2
214.039
MFCD00004171
YPJUNDFVDDCYIH-UHFFFAOYSA-N
heptafluorobutyric acid,perfluorobutyric acid,heptafluorobutanoic acid,perfluorobutanoic acid,butanoic acid, heptafluoro,heptafluoro-1-butanoic acid,perfluoropropanecarboxylic acid,butyric acid, heptafluoro,kyselina heptafluormaselna,kyselina heptafluormaselna czech
9777
CHEBI:39426
2,2,3,3,4,4,4-heptafluorobutanoic acid
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
Quantity 100g
Boiling Point 120°C
Color Yellow
Melting Point -20°C
UN Number 3265
Formula Weight 214.04
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Heptafluorobutyric Acid

EINECSNumber : (2)-1182

RTECSNumber : ET4025000

TSCA : Yes