Alfa Aesar™ Heptanal, 97%

Manufacturer: Alfa Aesar™ B23830AE

Quantity 100mL

 View more versions of this product

Catalog No. AAB23830AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 111-71-7
    Molecular Formula CH3(CH2)5CHO
    Formula Weight 114.19
    MDL Number MFCD00007028
    UN Number UN3056
    Beilstein 1560236
    Merck Index 14,4658
    Synonym Heptaldehyde
    Chemical Name or Material Heptanal

    Specifications

    Formula Weight 114.19
    Physical Form Liquid
    Assay Percent Range 0.97
    Storage Note 1 Hygroscopic
    TSCA TSCA
    Molecular Formula CH3(CH2)5CHO
    CAS 111-71-7
    UN Number UN3056
    Merck Index 14,4658
    Synonym Heptaldehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-HEPTALDEHYDE
    RTECS Number MI6900000
    Flash Point 35°C (95°F)
    Odor Unpleasant
    Melting Point −43°C
    Quantity 100mL
    MDL Number MFCD00007028
    Beilstein 1560236
    Boiling Point 152° to 154°C
    Chemical Name or Material Heptanal
    Assay 97%

    Description

    Applications
    Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

    Solubility
    Soluble in water 1.3 g/L.

    Notes
    Hygroscopic. Store away from oxidizing agents, air.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Hygroscopic

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit