Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Supplier:  MilliporeSigma Supelco 5173050ML

 View more versions of this product

Catalog No. 11-101-8189


Add to Cart

Description

Description

Petroleum
Specifications

Specifications

50 mL
Analytical Standard
MFCD00009544
≥99.8% (GC)
433°F
3.5 (vs. air)
-91°C
22°C
Neat
142-82-5
CH3(CH2)5 CH3
100.20
Analytical Standard
83 mmHg (37.7°C)
0.684 g/mL (at 25°C)
98°C
Insoluble in water
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : MI7700000

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit