Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane (Technical), Fisher Chemical™

Catalog Number p-216021
Click to view available options
Quantity:
20 L
200 L
Packaging:
Steel Drum
Steel Pail
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
Melting Point -91°C
Color Colorless
Density 0.683g/cm³
Boiling Point 98°C
Quantity 20 L
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Vapor Pressure 48mbar at 20°C
Viscosity 0.4 mPa/s at 20°C
UN Number 1206
Formula Weight 100.2
Grade Technical
Chemical Name or Material Heptane
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit