Alfa Aesar™ Heptanenitrile, 98%

Manufacturer: Alfa Aesar™ A1232606

Quantity 5g

 View more versions of this product

Catalog No. AAA1232606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 629-08-3
    Molecular Formula CH3(CH2)5CN
    Formula Weight 111.19
    MDL Number MFCD00039503
    UN Number UN1992
    Beilstein 1740405
    Synonym Enanthonitrile; Heptanonitrile
    Chemical Name or Material Heptanenitrile

    Specifications

    Formula Weight 111.19
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H331-H226-H302-H312
    Toxic if inhaled.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    TSCA TSCA
    Beilstein 1740405
    Synonym Enanthonitrile; Heptanonitrile
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Flash Point 58°C (136°F)
    MDL Number MFCD00039503
    CAS 629-08-3
    UN Number UN1992
    Health Hazard 1 Danger
    Boiling Point 186° to 187°C
    Quantity 5g
    Molecular Formula CH3(CH2)5CN
    Melting Point −64°C
    Chemical Name or Material Heptanenitrile
    Assay 98%

    Description

    Applications
    The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Strong reducing agents, Strong acids, Strong bases.

    GHS H Statement
    H331-H226-H302-H312
    Toxic if inhaled.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit