Alfa Aesar™ Heptanophenone, 98+%

Manufacturer: Alfa Aesar™ A1049118

Quantity 50g

 View more versions of this product

Catalog No. AAA1049118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1671-75-6
    Molecular Formula C13H18O
    Formula Weight 190.29
    MDL Number MFCD00009539
    Beilstein 1865692
    Synonym n-Hexyl phenyl ketone
    Chemical Name or Material Heptanophenone

    Specifications

    Formula Weight 190.29
    CAS 1671-75-6
    Beilstein 1865692
    Synonym n-Hexyl phenyl ketone
    Boiling Point 283°C
    Quantity 50g
    TSCA TSCA
    Molecular Formula C13H18O
    Flash Point >110°C (230°F)
    Melting Point ≈17°C
    MDL Number MFCD00009539
    Chemical Name or Material Heptanophenone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit