Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptyl Acetoacetate 95.0+%, TCI America™

Click to view available options
Quantity:
25mL
42598-96-9
C11H20O3
200.278
MFCD00027313
YAVJSVDUZGIQPQ-UHFFFAOYSA-N
Acetoacetic Acid Heptyl Ester
95760
heptyl 3-oxobutanoate
CCCCCCCOC(=O)CC(=O)C
42598-96-9
C11H20O3
200.278
MFCD00027313
YAVJSVDUZGIQPQ-UHFFFAOYSA-N
Acetoacetic Acid Heptyl Ester
95760
heptyl 3-oxobutanoate
CCCCCCCOC(=O)CC(=O)C
Color Yellow
Quantity 25mL
Boiling Point 170°C
Formula Weight 200.28
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material Heptyl Acetoacetate

EINECSNumber : (2)-1505

TSCA : No