Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptyloxyethanol, 90%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
7409-44-1
C9H20O2
160.257
MFCD01727884
QJTOHHRPYAVWOO-UHFFFAOYSA-N
2-heptyloxy ethanol,ethanol, 2-heptyloxy,heptyl cellosolve,ethylene glycol monoheptyl ether,2-heptyloxyethan-1-ol,heptyloxyethanol,3-oxadecane-1-ol,2-1-heptyloxy-1-ethanol
23889
2-heptoxyethanol
CCCCCCCOCCO
7409-44-1
C9H20O2
160.257
MFCD01727884
QJTOHHRPYAVWOO-UHFFFAOYSA-N
2-heptyloxy ethanol,ethanol, 2-heptyloxy,heptyl cellosolve,ethylene glycol monoheptyl ether,2-heptyloxyethan-1-ol,heptyloxyethanol,3-oxadecane-1-ol,2-1-heptyloxy-1-ethanol
23889
2-heptoxyethanol
CCCCCCCOCCO
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Assay Percent Range 90%
Linear Formula H3C(CH2)6O(CH2)2OH
Formula Weight 160.26
Physical Form Liquid
Percent Purity ≥88%
Chemical Name or Material Heptyloxyethanol, 90%

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.