Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu Ab-1 Rabbit Polyclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Epredia™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia RB103P1ABX

 View more versions of this product

Catalog No. RB103P1ABX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Rat and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human HER-2/c-erbB-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

HER-2/c-erbB-2/neu Ab-1
Polyclonal
Unconjugated
Synthetic peptide derived from C-terminus of human c-erbB-2/HER-2 protein. This Sequence is identical in rat neu protein.
RUO
Primary
Concentrated
Immunoprecipitation, Western Blot
1mg/mL
Rabbit
500 μL
Oncogenesis
Human, Mouse, Non-human Primate
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit