Learn More
HER-2/c-erbB-2/neu Ab-8 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS325P1ABX
Description
c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein. The c-erbB-2 protein is overexpressed in a variety of carcinomas especially those of breast and ovary.Host Species: Mouse
Clone: e2-4001
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Cytoplasmic domain
Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein
Molecular Weight: 185 kDa
Positive Control: SKBR-3 cells, or T47D cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunoprecipitation (Native and Denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
HER-2/c-erbB-2/neu Ab-8 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Oncogenesis | |
Human, Mouse, Rat | |
IgG1 |
Immunoprecipitation | |
e2-4001 | |
Unconjugated | |
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.