Learn More
Epredia™ Lab Vision™ HER-2/c-erbB-2/neu Rabbit Monoclonal Antibody supernatant
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ HER-2/c-erbB-2/neu, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
$6768.00
Specifications
Antigen | HER-2/c-erbB-2/neu |
---|---|
Clone | EP1045Y |
Applications | Immunohistochemistry |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RM2112S
|
Epredia™
RM2112SA |
Supernatant | 1 mL |
Each for $6,768.00
|
|
||||
RM2112S0
|
Epredia™
RM2112S0A |
Supernatant | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
HER2 gene encodes a transmembrane glycoprotein with tyrosine kinase activity. It is closely related in structure to the epidermal growth factor receptor c-erbB-3/HER3/neu oncoprotein. The protein is detectable in a subset of breast and other carcinomas (urinary bladder, stomach, lung, endometrium and ovary). In the case of breast cancer, over expression determined by immunohistochemistry has been shown to be associated with poor prognosis and an indication to initiate trastuzumab/herceptin therapy.Host Species: Rabbit
Clone: EP1045Y
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: A synthetic peptide corresponding to the c-terminus of human Her2
Molecular Weight: 185 kDa
Positive Control: Breast carcinomas
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
HER-2/c-erbB-2/neu | |
Immunohistochemistry | |
Rabbit | |
Primary |
EP1045Y | |
Unconjugated | |
A synthetic peptide corresponding to the c-terminus of human Her2 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.