Learn More
Selleck Chemical LLCSupplier Diversity Partner HER2-Inhibitor-1 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S27525MG
HER2-Inhibitor-1 is an analog of ARRY-380. ARRY-380 is a potent and selective HER2 inhibitor with IC50 of 8 nM, equipotent against truncated p95-HER2, 500-fold more selective for HER2 versus EGFR.
Catalog No. 50-194-8075