Learn More
Thermo Scientific™ Heracell™ VIOS™ 160i CO2 Incubator with Cell Locker™ System
Protected chambers for your most sensitive cells.  A breakthrough in cell culture management that maximizes your sample security
Manufacturer:  Thermo Scientific™ 51032124
 View more versions of this product
Catalog No. 11-675-960
Add to cart
Description
Description
The Thermo Scientific™ Heracell™ VIOS 160i CO2 incubator with the Thermo Scientific™ Cell Locker™ system is an innovative solution of removable, protected chambers designed for improved culturing efficiency and security for sensitive cultures. Six individual, autoclavable polycarbonate chambers divide the incubator chamber, isolating individual cell types or projects. Each Cell Locker chamber has dual 0.2µm membrane filters that permit air circulation but exclude microbial contaminants. When a Cell Locker chamber is opened, the others remain stable, preserving the in vitro environment.
Specifications
Specifications
Electropolished stainless steel
IR180SI
Covered integrated water reservoir
Ambient +3° to 55°C
CE, CSA
5.8 cu.ft., 165L per chamber
34.6 x 25.1 x 35.4 in. (88 x 63.7 x 90cm)
183 lb, 83 kg
1 to 20%
Optional 1-21% or 5-90%
≥93% @ 37°C
USB, and optional 4-20mA
22.7 x 18.5 x 23.9 in. (57.6 x 47 x 60.7cm)
120V 50/60Hz
Single 165L stainless steel chamber configured for Cell Locker system, (Cell Locker chambers ordered separately)