Learn More
Thermo Scientific™ Heracell™ VIOS 160i Tri-Gas CO2 Incubators, stainless-steel, with 1-21% O2 control PROMO
Achieve your next breakthrough. This CO2 incubator delivers the performance reliability, ease of operation, and value required to support a range of culturing needs.
$8945.00 - $9888.00
Specifications
Capacity (English) 5.8. cu. ft.
Capacity (English) Shelf 22/66 lb.
Capacity (Metric) 165 L
Capacity (Metric) Shelf 10/30 kg
Certifications/Compliance CE, CSA
View More Specs
Products 2
Catalog Number Mfr. No. Description CO2 Sensor Technology Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Description CO2 Sensor Technology Electrical Requirements Price Quantity & Availability  
13-998-213PRO View Documents Thermo Scientific™
51030403PRO
Single Tri-gas 165L incubator, 1-21% TC Sensor 120 V, 50/60 Hz Each for $8,945.00
Add to cart
13-998-132PRO View Documents Thermo Scientific™
51030405PRO
Single Tri-gas 165L incubator, 1-21% IR Sensor 120 V, 50/60 Hz Each for $9,888.00
Add to cart
Description
Description
Direct heat chamber
  • 165L (5.8 cu.ft.) electropolished stainless steel chamber with compact footprint, easily stackable
  • Standard right hand door swing, field reversible by qualified service support
THRIVE active airflow 
  • In-chamber fan gently distributes clean, humidified air throughout the chamber ensuring  homogeneous conditions
  • Fast recovery of all parameters in 10 minutes or less for stable culturing conditions
  • Auto-stop function disables fan operation during door openings to minimize air exchange
ISO Class 5 HEPA filtration
  • In-chamber HEPA continuously filters the entire chamber air volume every 60 seconds
  • ISO Class 5 clean room air quality is achieved in the culture chamber within 5 minutes
  • Continuous protection minimizes airborne contaminants settling on interior surfaces
Covered Integrated humidity reservoir
  • Directly heated water reservoir provides stable high relative humidity levels, preventing culture desiccation
  • Cover limits particles and spilled media from settling into the reservoir, keeping water clean
  • Separating water provides a condensation free incubation chamber, limiting areas for contaminants to breed
  • 5X faster humidity recovery than traditional water pan designs
  • Easy water handling: humidity reservoir may be filled or drained without removing shelves or cultures
  • Water level sensor indicates when a refill is needed
Steri-Run on-demand high temperature sterilization cycle
  • Independently validated overnight cycle ensures complete elimination of all biological contaminants
  • 180°C cycle designed for uniform 12-log sterilization on all chamber surfaces
  • Simplifies cleaning and eliminates the need for separate autoclaving and reassembly of components
Intuitive, interactive iCAN touchscreen control  
  • Bright LED screen to monitor all incubator interactions
  • Provides visibility to changes in culture environment with performance trend graphing, error and usage logs
  • Convenient on-screen user prompts and reminders for easy navigation
  • Selectable languages simplify operation: English, Spanish, German, French, Italian, Japanese and Mandarin
In situ sensors and probes designed for precise control
  • Dual temperature probes for over temperature protection with operational back up
  • Thermal conductivity (TC) CO2 sensor with rH compensation designed for fast recovery and stable operation
Specifications
Specifications
5.8. cu. ft.
165 L
CE, CSA
Electropolished Stainless Steel
22.7 in.
22/66 lb.
10/30 kg
Powder Coated Cold Rolled Steel
USB, and optional 4-20mA
576 mm