Learn More
Thermo Scientific™ HeraSafe 2030i 6ft NSF-CTS Series, Factory acceptance testing documentation, IQ/OQ Field Service
HeraSafe 2030i-CTS Series, Complete Validation Package including Compliance Binder, IQ/OQ Documents supporting a timely and rapid validation process
$19646.00 - $23083.00
Products 2
Catalog Number Mfr. No. Description Dimensions (L x W x H) Exterior Price Quantity & Availability  
Catalog Number Mfr. No. Description Dimensions (L x W x H) Exterior Price Quantity & Availability  
09-034-000
View Documents
Thermo Scientific™
51033638
Herasafe 2030 i, 4 ft., with 10 " sash opening, with indented single piece work tray. 31.5 x 51.2 x 60.5
Each for $19,646.00
Add to cart
09-034-002
View Documents
Thermo Scientific™
51033639
Herasafe 2030 i, 6 ft., with 10 " sash opening and indented single piece work tray. 31.5 x 74.8 x 60.5
Each for $23,083.00
Add to cart
Description
Description
HeraSafe 2030i-CTS Series, Complete Validation Package including Compliance Binder, IQ/OQ Documents supporting a timely and rapid validation process. The Herasafe 2030i-CTS Series BSC is designed to deliver performance with superior precision for demanding cell culture applications and other sensitive work requiring assured safety for product, personnel and environment, especially for bio pharma and advanced research initiatives.