Learn More
Thermo Scientific™ Heratherm™ General Protocol Ovens: PROMO
Perform routine applications simply and safely with Thermo Scientific™ Heratherm™ General Protocol Ovens
$1446.00 - $2344.00
Specifications
Timer ON / OFF
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Type Electrical Requirements Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Type Electrical Requirements Dimensions (L x W x H) Price Quantity & Availability  
51-028-112HSD View Documents Thermo Scientific™
51028112SD
65 L Gravity Convection 120 V 60 Hz 22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm) Each for $1,446.00
Add to cart
51-028-121HSD View Documents Thermo Scientific™
51028121SD
66 L Mechanical Convection 120 V 60 Hz 22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm) Each for $1,687.00
Add to cart
51-028-872HSD View Documents Thermo Scientific™
51028872SD
105 L Gravity Convection 120 V 60 Hz 22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm) Each for $1,671.00
Add to cart
51-028-873HSD View Documents Thermo Scientific™
51028873SD
104 L Mechanical Convection 120 V 60 Hz 22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm) Each for $2,009.00
Add to cart
51-028-874HSD View Documents Thermo Scientific™
51028874SD
176 L Gravity Convection 120 V 60 Hz 29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm) Each for $2,005.00
Add to cart
51-028-875HSD View Documents Thermo Scientific™
51028875SD
179 L Mechanical Convection 120 V 60 Hz 29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm) Each for $2,344.00
Add to cart
Description
Description
Perform routine applications simply and safely with Thermo Scientific™ Heratherm™ General Protocol Ovens, which provide an ideal heating and drying solution for research, clinical or industrial needs.
Heratherm General Protocol Ovens provide a choice of gravity convection for gentle drying and heating, or mechanical convection for faster drying and heating, and superior temperature uniformity. Gravity convection units are designed to protect delicate samples while offering temperature uniformity of ±4°C or better, and temperature stability of at least ±0.4°C at 150°C. Mechanical convection models provide temperature uniformity of ±4.0°C or better, and temperature stability of at least ±0.4°C at 150°C.
  • Five sizes (60, 100, 180, 400, or 750L)
  • Operating temperatures from 50° to 250°C
  • Automatic over-temperature alarm system to protect samples
  • Low overall energy consumption
  • Built in timer
  • Excellent footprint/volume ratio
  • Corrosion-resistant galvanized steel chambers with rounded corners for easy cleaning
  • Large, easy to read vacuum fluorescent display
  • Easy to use, microprocessor-controlled touch button operation
  • Easy to access—doors can be opened to over 180°C
Specifications
Specifications
ON / OFF
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.