Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549442

Catalog No. PA549442

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable E3 ubiquitin-protein ligase involved in either protein trafficking or in the distribution of cellular structures. Required for spermatozoon maturation and fertility, and for the removal of the cytoplasmic droplet of the spermatozoon. E3 ubiquitin-protein ligases accept ubiquitin from an E2 ubiquitin- conjugating enzyme in the form of a thioester and then directly transfer it to targeted substrates.
Specifications

Specifications

HERC4
Polyclonal
PBS with 0.09% sodium azide
Q5GLZ8
Probable E3 ubiquitin-protein ligase HERC4, 632-, HECT domain and RCC1-like domain-containing protein 4, HERC4, KIAA1593
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26091
Western Blot
Unconjugated
HERC4
Liquid
HERC4
KLH conjugated synthetic peptide between 445-479 amino acids from the Central region of human HERC4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit