Learn More
HERC5 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 226921AP
Catalog No. 50-173-3926
Add to cart
Description
Description
HERC5 is a probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. This gene is a member of the HERC family of ubiquitin ligases and encodes a protein with a HECT domain and five RCC1 repeats. Pro-inflammatory cytokines upregulate expression of this gene in endothelial cells. The protein localizes to the cytoplasm and perinuclear region and functions as an interferon-induced E3 protein ligase that mediates ISGylation of protein targets. The gene lies in a cluster of HERC family genes on chromosome 4.
Specifications
Specifications
HERC5
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UII4
CEB1, CEBP1, Cyclin E binding protein 1, E3 ISG15 protein ligase HERC5, hect domain and RLD 5, HERC5
Rabbit
IgG
150 μL
-20°C
Primary
51191
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
HERC5
Liquid
HERC5
HERC5 Fusion Protein Ag18449
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human