Learn More
BioworldSupplier Diversity Partner Herpes Simplex Virus-1 gG (Recombinant), 100 μg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220601851
Recombinant Herpes Simplex Virus-1 glycoprotein G (HSV-1-gG) Source: Saccharomyces Cerevisiae Description : Recombinant HSV-1 gG contains the immunodominant regions of glycoprotein G, amino acids (84-175). HSV-1-gG is purified by proprietary chromatographic techniques. Formulation: 1mg/ml in 20mM sodium phosphate pH-7.5, 1mM EDTA and 6M urea. Purity: Greater than 95.0% as determined by: (a) 10% PAGE coomassie staining. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Please avoid freeze-thaw cycles. Stability: Five years frozen. One month in solution at room temperature Specificity: Immunoreactive with sera of HSV-infected individuals. Applications: 1. ELISA 2. Western blots 3. HSV Detection. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-285
Due to product restrictions, please to purchase or view availability for this product.