Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 (HSV1) Mouse anti-Virus, Clone: 10A3, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Herpes Simplex Virus 1 (HSV1)
Clone 10A3
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123088
View Documents
Novus Biologicals
NBP30780720UG
20 μg
Each for $138.37
N/A
Due to product restrictions, please to purchase or view availability for this product.
NB123087
View Documents
Novus Biologicals
NBP30780710UG
100 μg
Each for $352.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Herpes Simplex Virus 1 (HSV1) Monoclonal antibody specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Herpes Simplex Virus 1 (HSV1)
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
Detergent-solubilized herpes simples virus (HSV) type 1 infected cells
Monoclonal
Protein A or G purified
10A3
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Virus
Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit