Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 (HSV1) Mouse anti-Virus, Clone: HSVI/2045, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Herpes Simplex Virus 1 (HSV1)
Clone HSVI/2045
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122861
View Documents
Novus Biologicals
NBP30769410UG
100 μg
Each for $352.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
NB122862
View Documents
Novus Biologicals
NBP30769420UG
20 μg
Each for $138.37
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Herpes Simplex Virus 1 (HSV1) Monoclonal antibody specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Western Blot
Specifications

Specifications

Herpes Simplex Virus 1 (HSV1)
0.2 mg/ml
Western Blot
Purified
RUO
10 mM PBS with 0.05% BSA
Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex
Monoclonal
Protein A or G purified
HSVI/2045
Western Blot 1-2 ug/ml
Unconjugated
Mouse
Virus
Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit