Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 (HSV1) Rabbit anti-Virus, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen Herpes Simplex Virus 1 (HSV1)
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122110
View Documents
Novus Biologicals
NBP30731820UG
20 μg
Each for $138.37
N/A
Due to product restrictions, please to purchase or view availability for this product.
NB122109
View Documents
Novus Biologicals
NBP30731810UG
100 μg
Each for $352.89
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Herpes Simplex Virus 1 (HSV1) Polyclonal antibody specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Herpes Simplex Virus 1 (HSV1)
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Virus
Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2
IgG
Primary
Store at 4°C.
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
Detergent-solubilized herpes simplex virus (HSV) type 1 infected cells
Polyclonal
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit