Learn More
Invitrogen™ Herpes Simplex Virus Type 1/2 gB Monoclonal Antibody (T111)

Description
This antibody recognizes gB antigen of both HSV1 and HSV2 (Herpes Simplex Virus type 1 and 2), a dsDNA virus, member of Herpesviridae family.
Specifications
Specifications
| Antigen | Herpes Simplex Virus Type 1/2 gB |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | T111 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15 mM sodium azide; pH 7.4 |
| Gene Alias | Herpes Simplex 1; Herpes Simplex 2; Herpes Simplex I; Herpes Simplex II; HHV; HHV-2; HSV; HSV 1/2; HSV I/II; HSV Type 1; HSV Type 2; HSV Type II; HSV1; HSV2; HSV-2 |
| Host Species | Mouse |
| Immunogen | Exctract of HSV-1 infected VERO cells.. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.