Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes simplex virus type 1/2 ICP5 Mouse anti-Herpes simplex virus, Clone: HA018, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13576
Description
Mouse monoclonal antibody raised against herpes simplex virus type 1/2 (HSV-1/2) ICP5.
- High specificity
- High reactivity
Specifications
Herpes simplex virus type 1/2 ICP5 | |
Monoclonal | |
Unconjugated | |
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user. | |
Mouse | |
RUO | |
This antibody reacts with ICP5 of herpes simplex virus type 1/2. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b λ |
ELISA, Immunofluorescence, Western Blot | |
HA018 | |
Mouse monoclonal antibody raised against herpes simplex virus type 1/2 (HSV-1/2) ICP5. | |
In PBS, pH 7.4. | |
100 μg | |
Primary | |
Herpes Simplex virus (HSV) | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction