Learn More
Herpes simplex virus type 1 gC Mouse anti-Herpes simplex virus, Clone: H1A022, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13577
Catalog No. 89-033-540
Add to cart
Description
Description
Mouse monoclonal antibody raised against herpes simplex virus type 1 (HSV-1) gC.
  • High specificity
  • High reactivity
Specifications
Specifications
Herpes simplex virus type 1 gC
H1A022
Mouse monoclonal antibody raised against herpes simplex virus type 1 (HSV-1) gC.
In PBS, pH 7.4.
Mouse
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Herpes Simplex Virus
ELISA, Immunofluorescence, Western Blot
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG2a, kappa
RUO
Primary
This antibody reacts with gC of herpes simplex virus type 1.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)