Learn More
Herpes simplex virus type 2 gG-2 Mouse anti-Herpes simplex virus, Clone: P1106, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13590
Catalog No. 89-033-553
Add to cart
Description
Description
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gG-2.
  • High specificity
  • High reactivity
Specifications
Specifications
Herpes simplex virus type 2 gG-2
P1106
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gG-2.
In PBS, pH 7.4.
Mouse
Protein G purification
RUO
Primary
This antibody reacts with gG-2 of herpes simplex virus type 2.
Immunofluorescence, Western Blot
Unconjugated
Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG1, kappa
500 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Herpes Simplex Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)