Learn More
Herpes simplex virus type 2 ICP8 Mouse anti-Herpes simplex virus, Clone: H2A024, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13592
Catalog No. 89-033-555
Add to cart
Description
Description
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) ICP8.
  • High specificity
  • High reactivity
Specifications
Specifications
Herpes simplex virus type 2 ICP8
H2A024
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) ICP8.
In PBS, pH 7.4.
Mouse
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Herpes Simplex Virus
ELISA, Immunofluorescence, Western Blot
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG2a, kappa
RUO
Primary
This antibody reacts with ICP8 of herpes simplex virus type 2.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)