Learn More
Herpes simplex virus type 2 Sheep anti-Herpes simplex virus, Polyclonal, Abnova™
Sheep Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29862
Catalog No. 89-033-707
Add to cart
Description
Description
Sheep polyclonal antibody raised against herpes simplex virus type 2 (HSV-2).
  • High specificity
  • High reactivity
Specifications
Specifications
Herpes simplex virus type 2
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Herpes simplex virus type 2 strain G.
RUO
Primary
Herpes Simplex Virus
ELISA, Immunofluorescence, Western Blot
Sheep polyclonal antibody raised against herpes simplex virus type 2 (HSV-2).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Sheep
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)