Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Herpes Simplex Virus Type 2 Strain Culture Fluid

Infectious virus preparation designed to meet research and product development needs

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration >105U/mL
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
View More Specs

Alert:

Contains live and infectious organisms.


Products 21
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22-157-552
View Documents Promotion Details
ZeptoMetrix
0810006CFN
Herpes Simplex Virus Type 2 Isolate 1 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-739
View Documents Promotion Details
ZeptoMetrix
0810203CF
Herpes Simplex Virus Type 2 Isolate 2 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-740
View Documents Promotion Details
ZeptoMetrix
0810204CF
Herpes Simplex Virus Type 2 Isolate 3 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-741
View Documents Promotion Details
ZeptoMetrix
0810205CF
Herpes Simplex Virus Type 2 Isolate 4 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-742
View Documents Promotion Details
ZeptoMetrix
0810206CF
Herpes Simplex Virus Type 2 Isolate 5 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-747
View Documents Promotion Details
ZeptoMetrix
0810211CF
Herpes Simplex Virus Type 2 Isolate 10 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-745
View Documents Promotion Details
ZeptoMetrix
0810209CF
Herpes Simplex Virus Type 2 Isolate 8 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-746
View Documents Promotion Details
ZeptoMetrix
0810210CF
Herpes Simplex Virus Type 2 Isolate 9 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-744
View Documents Promotion Details
ZeptoMetrix
0810208CF
Herpes Simplex Virus Type 2 Isolate 7 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-743
View Documents Promotion Details
ZeptoMetrix
0810207CF
Herpes Simplex Virus Type 2 Isolate 6 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-748
View Documents Promotion Details
ZeptoMetrix
0810212CF
Herpes Simplex Virus Type 2 Isolate 11 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-749
View Documents Promotion Details
ZeptoMetrix
0810213CF
Herpes Simplex Virus Type 2 Isolate 12 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-750
View Documents Promotion Details
ZeptoMetrix
0810214CF
Herpes Simplex Virus Type 2 Isolate 13 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-752
View Documents Promotion Details
ZeptoMetrix
0810216CF
Herpes Simplex Virus Type 2 Isolate 15 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-751
View Documents Promotion Details
ZeptoMetrix
0810215CF
Herpes Simplex Virus Type 2 Isolate 14 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-753
View Documents Promotion Details
ZeptoMetrix
0810217CF
Herpes Simplex Virus Type 2 Isolate 16 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-754
View Documents Promotion Details
ZeptoMetrix
0810218CF
Herpes Simplex Virus Type 2 Isolate 17 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-756
View Documents Promotion Details
ZeptoMetrix
0810220CF
Herpes Simplex Virus Type 2 Isolate 19 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-757
View Documents Promotion Details
ZeptoMetrix
0810221CF
Herpes Simplex Virus Type 2 Isolate 20 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-755
View Documents Promotion Details
ZeptoMetrix
0810219CF
Herpes Simplex Virus Type 2 Isolate 18 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-758
View Documents Promotion Details
ZeptoMetrix
0810222CF
Herpes Simplex Virus Type 2 Isolate 21 Infectious Culture Fluid
Each for $354.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Human Herpes viruses are enveloped icosahedral viruses 150 to 200nm in diameter that contain a nucleocapsid and double-stranded linear DNA.

  • Consist of virus, cells and media taken directly from tissue culture flask
  • Propagated in Vero cell line
  • Biosafety Level 2 organism
  • Available in 1.0mL aliquots
  • Each lot assayed for its Tissue Culture Infective Dose (TCID50)
  • Titer:>105U/mL

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Nucleic acid/molecular testing, Limit of Detection (LOD) studies, Cross-reactivity studies, Other viral-based assays

Specifications

Specifications

2
>105U/mL
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
1 mL
ISO, 13485
Solution
Vial
Safety and Handling

Safety and Handling

ShelfLife : 1 Year

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit