Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERPUD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $299.00

Specifications

Antigen HERPUD2
Immunogen Synthetic peptides corresponding to HERPUD2(HERPUD family member 2) The peptide sequence was selected from the N terminal of HERPUD2 (NP_071768). Peptide sequence MDQSGMEIPVTLIIKAPNQKYSDQTISCFLNWTVGKLKTHLSNVYPSKPL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15972420 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15972420UL
20ul Each for $149.00
Add to cart
 
NBP159724 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159724
50ul Each for $299.00
Add to cart
 
Description

Description

HERPUD2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

HERPUD2
Rabbit
Polyclonal
Unconjugated
64224
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to HERPUD2(HERPUD family member 2) The peptide sequence was selected from the N terminal of HERPUD2 (NP_071768). Peptide sequence MDQSGMEIPVTLIIKAPNQKYSDQTISCFLNWTVGKLKTHLSNVYPSKPL.
Primary
IgG
endoplasmic reticulum stress-inducible, ubiquitin-likedomain member 2, FLJ22313, FLJ31032, HERPUD family member 2, homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domainmember 2 protein
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit