Learn More
Seracare Life Sciences IncSupplier Diversity Partner HERRING SPERM DNA  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 59200003
Biotechnology grade, sheared and denatured, Herring Sperm DNA is used as a blocking agent in prehybridization and hybridization procedures to minimize nonspecific binding of hybridization probe to membranes and in situ.
Catalog No. 50-675-44