Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HERV-FRD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544470
Description
Peptide sequence: TGSPYSTNCW LCTSSSTETP GTAYPASPRE WTSIEAELHI SYRWDPNLKG Sequence homology: Human: 100%.
Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. This endogenous envelope protein has retained its original fusogenic properties. HERV-FRD can make pseudotypes with MLV, HIV-1 or SIV-1 virions and confer infectivity.Human endogenous retroviruses (HERVs) make up approximately 8% of the human genome. Although most HERVs are nonfunctional, the HERV-W (ERVWE1; MIM 604659) and HERV-FRD envelope (env) proteins can induce cell-cell fusion when expressed in cells possessing appropriate receptors (Blaise et al., 2003 [PubMed 14557543]).[supplied by OMIM].
Specifications
HERV-FRD | |
Polyclonal | |
Unconjugated | |
ERVFRD-1 | |
Endogenous retrovirus group FRD member 1; endogenous retrovirus group FRD, member 1; envelope glycoprotein syncytin-A; envelope polyprotein; envFRD; ERVFRD-1; ERVFRDE1; FLJ41944; FLJ90611; GLLL6191; Gm52; HERV-FRD; HERV-FRD provirus ancestral Env polyprotein; HERV-FRD_6p24.1 provirus ancestral Env polyprotein; HERV-W/FRD; MGC87585; SU; Surface protein; Syna; Synb; syncytin 2; syncytin a; Syncytin2; syncytin-2; TM; Transmembrane protein; UNQ6191; UNQ6191/PRO20218 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
405754 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P60508 | |
ERVFRD-1 | |
Synthetic peptide directed towards the N-terminal of human HERV-FRD (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction