Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA561626

Catalog No. PIPA561626

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Immunogen sequence: RARINVSLEQ LKSLLEKHYS HQIRKRKLEK ADHighest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.
Specifications

Specifications

HES3
Polyclonal
Unconjugated
HES3
Liquid
Rabbit
IgG
100μL
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles.
Primary
390992
Immunohistochemistry
0.2mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5TGS1
HES3
Recombinant protein corresponding to Human HES3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only