Alfa Aesar™ Heterogeneous Catalyst Kit, 40 sample
Manufacturer: Alfa Aesar™ H37163KT
 View more versions of this product
Catalog No. AAH37163KT
  Check Availability
Add to Cart
    Specifications
    Quantity 1 Kit
    Product Type Catalyst
    Synonym Global Heterogeneous Catalyst Kit, 40 sample
    Excellent tools for screening and research projects. Provides instant availability to 40 catalyst types with varying characteristics and metal distribution. The kit is supplied with an Applications Table that allows for easy identification of prime candidates for catalytic processes.
  • Description & Specifications
Related Products
Product Content Feedback