Alfa Aesar™ Hexa(acetato)-mu-oxotris(aqua)trirhodium(III) acetate

Manufacturer: Alfa Aesar™ 43004-03

Quantity 1g

 View more versions of this product

Catalog No. AA4300403

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 42204-14-8
    Molecular Formula C14H27O20Rh3
    Formula Weight 824.07
    MDL Number MFCD01074923
    Synonym Rhodium(III) acetate
    Chemical Name or Material Hexa (acetato)-mu-oxotris (aqua)trirhodium (III) acetate

    Specifications

    Formula Weight 824.07
    Synonym Rhodium(III) acetate
    Health Hazard 2 GHS H Statement
    H341-H319
    Suspected of causing genetic defects.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P281-P305+P351+P338-P308+P313-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for sever
    Quantity 1g
    MDL Number MFCD01074923
    Solubility Information Soluble in water, acetic acid
    Health Hazard 1 Warning
    Color Brown
    Physical Form Powder
    Molecular Formula C14H27O20Rh3
    CAS 42204-14-8
    Chemical Name or Material Hexa (acetato)-mu-oxotris (aqua)trirhodium (III) acetate

    GHS H Statement
    H341-H319
    Suspected of causing genetic defects.
    Causes serious eye irritation.

    GHS P Statement
    P280-P281-P305+P351+P338-P308+P313-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for sever

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit