Hexaamminecobalt(III) chloride, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar 1376922

Quantity 100g

 View more versions of this product

Catalog No. AA1376922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10534-89-1
    Physical Form Crystalline
    Assay Percent Range 97%
    Molecular Formula [Co(NH3)6]Cl3
    Formula Weight 267.48
    MDL Number MFCD00036304
    Chemical Name or Material Hexaamminecobalt(III) chloride

    Specifications

    Formula Weight 267.48
    Quantity 100g
    Color Orange
    EINECS Number 234-103-9
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 97%
    Molecular Formula [Co(NH3)6]Cl3
    Appearance Orange powder
    MDL Number MFCD00036304
    CAS 10534-89-1
    Health Hazard 1 H351-H317; Suspected of causing cancer. May cause an allergic skin reaction.
    Chemical Name or Material Hexaamminecobalt(III) chloride

    H351-H317; Suspected of causing cancer. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit