Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hexaamminecobalt(III) nitrate

Manufacturer:  Alfa Aesar™ 2255514

Quantity 25g

 View more versions of this product

Catalog No. AA2255514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

10534-86-8
347.13
UN1477
[Co(NH3)6](NO3)3
MFCD00061421
Hexaamminecobalt(III) nitrate

Specifications

25g
Powder
347.13
UN1477
Hexaamminecobalt(III) nitrate
10534-86-8
[Co(NH3)6](NO3)3
MFCD00061421
1.804g/mL

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H351-H317
May intensify fire
oxidizer.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit