Alfa Aesar™ Hexaamminecobalt(III) nitrate

Manufacturer: Alfa Aesar™ 2255506

Quantity 5g

 View more versions of this product

Catalog No. AA2255506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10534-86-8
    Linear Formula [Co(NH3)6](NO3)3
    Formula Weight 347.13
    MDL Number MFCD00061421
    UN Number UN1477
    Chemical Name or Material Hexaamminecobalt(III) nitrate

    Specifications

    Formula Weight 347.13
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H272-H351-H317
    May intensify fire
    oxidizer.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Physical Form Powder
    MDL Number MFCD00061421
    TSCA TSCA
    Linear Formula [Co(NH3)6](NO3)3
    UN Number UN1477
    Health Hazard 1 Danger
    Density 1.804g/mL
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    Quantity 5g
    CAS 10534-86-8
    Chemical Name or Material Hexaamminecobalt(III) nitrate

    GHS H Statement
    H272-H351-H317
    May intensify fire
    oxidizer.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit