Alfa Aesar™ Hexaamminenickel(II) bromide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4448222

Quantity 100g

 View more versions of this product

Catalog No. AA4448222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13601-55-3
    Molecular Formula Ni(NH3)6Br2
    Formula Weight 320.68
    MDL Number MFCD04039935
    Chemical Name or Material Hexaamminenickel (II) bromide

    Specifications

    Formula Weight 320.68
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula Ni(NH3)6Br2
    Health Hazard 1 Danger
    MDL Number MFCD04039935
    CAS 13601-55-3
    Health Hazard 2 GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.
    Physical Form Crystalline
    Quantity 100g
    Chemical Name or Material Hexaamminenickel (II) bromide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit