Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hexaammineruthenium(II) chloride, 99.9%

Click to view available options
Quantity:
0.25g
1g
15305-72-3
ClH12N6Ru
232.66
MFCD00011477
MHYZHXDLILNYTQ-UHFFFAOYSA-M
triton rw 30,azane;ruthenium 2+ ;dichloride,ruthenium iii chloride hexaammoniate,hexaammineruthenium ii chloride
15305-72-3
ClH12N6Ru
232.66
MFCD00011477
MHYZHXDLILNYTQ-UHFFFAOYSA-M
triton rw 30,azane;ruthenium 2+ ;dichloride,ruthenium iii chloride hexaammoniate,hexaammineruthenium ii chloride
Melting Point >300°C
Quantity 1g
Odor Odorless
Formula Weight 274.16
Percent Purity 99.9%
Chemical Name or Material Hexaammineruthenium(II) chloride

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C