Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hexaammineruthenium(III) chloride, Ru 32.1% min

Manufacturer:  Alfa Aesar™ 1051101

Quantity 0.25g

 View more versions of this product

Catalog No. AA1051101


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14282-91-8
MFCD00011478
Hexaammineruthenium(III) chloride
309.61
UN3288

Specifications

Ru 32.1% min.
0.25g
Powder
MFCD00011478
Hexaammineruthenium(III) chloride
Pale yellow
14282-91-8
309.61
UN3288

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : VM2651530

storageNote1 : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit