Alfa Aesar™ Hexacarbonylmolybdenum, 98%

Manufacturer: Alfa Aesar™ 1305714

Quantity 25g

 View more versions of this product

Catalog No. AA1305714

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13939-06-5
    Molecular Formula C6MoO6
    Linear Formula Mo(CO)6
    Formula Weight 264
    MDL Number MFCD00003466
    UN Number UN3466
    Synonym Molybdenum hexacarbonyl; Molybdenum carbonyl
    Chemical Name or Material Hexacarbonylmolybdenum, Up to 1% Cr

    Specifications

    Formula Weight 264
    Physical Form Crystalline
    Assay Percent Range 98%
    Molecular Formula C6MoO6
    Health Hazard 3 GHS P Statement
    P280f-P304+P340-P310-P302+P352
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    CAS 13939-06-5
    Melting Point ≈150°C (dec.)
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    UN Number UN3466
    Synonym Molybdenum hexacarbonyl; Molybdenum carbonyl
    Health Hazard 1 Danger
    Linear Formula Mo(CO)6
    MDL Number MFCD00003466
    Density 1.96g/mL
    Quantity 25g
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.
    Color White
    Concentration May contain up to 1% Cr
    Chemical Name or Material Hexacarbonylmolybdenum, Up to 1% Cr
    Assay 98%

    GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.

    GHS P Statement
    P280f-P304+P340-P310-P302+P352
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit